missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate monohydrate, 95%, Thermo Scientific Chemicals

Product Code. 11358708
Click to view available options
Quantity:
500 g
2 kg

Potassium phosphate monohydrate acts as a reagent and buffer used in molecular biology, biochemistry, and chromatography. It is utilized to study the effects of freezing and thawing of proteins sensitive to conformational changes. It is also used in the extraction of keratohyalin protein from bovine tissue. Further, it is used as a food additive, emulsifier, foaming agent and fertilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27176-10-9
Molecular Formula H2K3O5P
Molecular Weight (g/mol) 230.28
MDL Number MFCD00150410
InChI Key RMNIZOOYFMNEJJ-UHFFFAOYSA-K
Synonym tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7
PubChem CID 22031701
IUPAC Name tripotassium;phosphate;hydrate
SMILES O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Color White
Physical Form Granules
Quantity 500 g
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 230.28 (212.27 Anhydrous)
Odor Odorless
Appearance White granules
Density 2.564
Chemical Name or Material Potassium phosphate monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'