missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate, tribasic, 97%, pure, anhydrous
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
25g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1340.0°C |
| Color | White |
| Physical Form | Powder, Granules or Chunks |
| Quantity | 10 kg |
| Assay Percent Range | 97% |
| Linear Formula | K3PO4 |
| Merck Index | 15,7781 |
| Solubility Information | Solubility in water: 508g/L (25°C). Other solubilities: insoluble in alcohol |
| Formula Weight | 212.27 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
Product Identifier
- Potassium phosphate tribasic
- Danger
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction