missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium polysulfide, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 200.0°C to 250.0°C |
Quantity | 250 g |
Linear Formula | (K2S)n |
Solubility Information | Solubility in water: 500g/l (20°C) |
IUPAC Name | potassium sulfane |
Grade | Pure |
Packaging | Plastic bottle |
Chemical Name or Material | Potassium polysulfide |
Safety and Handling
Product Identifier
- Potassium polysulfide
- Danger
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 1 B
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- EUH031-Contact with acids liberates toxic gas.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction