missing translation for 'onlineSavingsMsg'
Learn More

Potassium selenocyanate, 99%, Thermo Scientific Chemicals

Product Code. 10114030
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3425-46-5
Molecular Formula CKNSe
Molecular Weight (g/mol) 144.08
MDL Number MFCD00011366
InChI Key KYEKHFSRAXRJBR-UHFFFAOYSA-M
Synonym potassium selenocyanate, selenocyanic acid, potassium salt, potassium selenoisocyanate, ccris 7063, potassium ion selenocyanate, potassium selenocyanate 10g
PubChem CID 76960
IUPAC Name potassium;selenocyanate
SMILES C(#N)[Se-].[K+]

Specifications

Melting Point 152.0°C to 158.0°C
Color Orange to Red
Physical Form Crystalline Solid, Powder, Crystals and/or Chunks
Quantity 50 g
Assay Percent Range 98.5 % min. (Argentometry)
Linear Formula KSeCN
Fieser 06,487; 08,417; 13,186; 08,417; 15,274
Solubility Information Solubility in water: soluble
Formula Weight 144.08
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Potassium selenocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium selenocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium selenocyanate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.