missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium silver cyanide, 99.9% (metals basis), Ag 54.2%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Silver platingPotassium silver cyanide is used in silver plating and in the manufacturing of antiseptics. It is a versatile bridging ligand and finds use for multidimensional polymer construction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | KAg(CN)2 |
UN Number | UN1588 |
Merck Index | 14,7669 |
Solubility Information | Soluble in waterSoluble in water; Insoluble in acids. |
Formula Weight | 199.01 |
Percent Purity | 99.9% |
Concentration or Composition (by Analyte or Components) | Ag 54.2% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'