missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sodium tartrate tetrahydrate, ACS reagent, Thermo Scientific Chemicals
35.07 € - 266.53 €
Chemical Identifiers
CAS | 6381-59-5 |
---|---|
Molecular Formula | C4H4KNaO6·4H2O |
Molecular Weight (g/mol) | 282.22 |
InChI Key | VZOPRCCTKLAGPN-UHFFFAOYSA-L |
Synonym | potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j |
PubChem CID | 2724148 |
IUPAC Name | potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
SMILES | C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12675067
|
Thermo Scientific Acros
447221000 |
100 g |
35.07 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12685067
|
Thermo Scientific Acros
447225000 |
500 g |
70.25 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12643600
|
Thermo Scientific Acros
447220025 |
2.5 kg |
266.53 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6381-59-5 | |
282.22 | |
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
C4H4KNaO6·4H2O | |
VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
2724148 | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
Specifications
70.0°C to 80.0°C | |
C4H4KNaO6·4H2O | |
15,779 | |
Solubility in water: 630g/L (20°C). Other solubilities: practically insoluble in alcohol | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] | |
282.22 | |
282.22 | |
Plastic bottle |
6381-59-5 | |
KO2CCH(OH)CH(OH)CO2Na·4H2O | |
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate | |
2724148 | |
ACS Reagent | |
Potassium sodium tartrate tetrahydrate |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title