missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sodium Tartrate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™
Potassium Sodium Tartrate Tetrahydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 6381-59-5, White, Melting Point: 70 deg.C, Molecular Formula: C4H12KNaO10, Molecular Weight: 282.218, Packaging: Plastic powder jar, pH: 5.5 to 8.5
140.00 € - 178.00 €
Chemical Identifiers
CAS | 6381-59-5 |
---|---|
Molecular Formula | C4H12KNaO10 |
Molecular Weight (g/mol) | 282.218 |
MDL Number | 150989 |
InChI Key | VZOPRCCTKLAGPN-UHFFFAOYSA-L |
Synonym | potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j |
PubChem CID | 2724148 |
IUPAC Name | potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
SMILES | C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10529200
|
Fisher Chemical
P/6880/53 |
500 g |
140.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10346240
|
Fisher Chemical
P/6880/60 |
1 kg |
178.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10161960
|
Fisher Chemical
P/6880/62 |
2.5 kg |
163.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium Sodium Tartrate Tetrahydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 6381-59-5, White, Melting Point: 70 deg.C, Molecular Formula: C4H12KNaO10, Molecular Weight: 282.218, Packaging: Plastic powder jar, pH: 5.5 to 8.5
Chemical Identifiers
6381-59-5 | |
282.218 | |
VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
2724148 | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
C4H12KNaO10 | |
150989 | |
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
Specifications
70°C | |
White | |
Powder | |
150989 | |
VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate | |
2724148 | |
Plastic powder jar |
6381-59-5 | |
5.5 to 8.5 | |
C4H12KNaO10 | |
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] | |
282.218 | |
282.22g/mol |