missing translation for 'onlineSavingsMsg'
Learn More

Potassium, solid, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11311398
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-09-7
Molecular Formula K
Molecular Weight (g/mol) 39.10
MDL Number MFCD00133776
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
Synonym kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
PubChem CID 5462222
ChEBI CHEBI:26216
IUPAC Name potassium
SMILES [K]

Specifications

Color Silver
Appearance Silver colored solid
Assay Percent Range 99.95%
Quantity 1 g
Odor Odorless
Chemical Name or Material Potassium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'