missing translation for 'onlineSavingsMsg'
Learn More

Potassium Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical

Brand:  Fisher Chemical J/8300/05


Product Code. 10244580

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

100mL, HDPE plastic bottle

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Colorless
Liquid
11409
1
100 mL
HDPE plastic bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium solution 10 000 ppm

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers