missing translation for 'onlineSavingsMsg'
Learn More

Potassium standard solution, for AAS, 1 mg/ml K in 2-5% HNO3

Product Code. 10022580 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 10022580

Brand: Thermo Scientific Acros 196205000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-09-7 | K | 39.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium standard solution
Name Note 1 mg/ml K in 2% HNO3
Packaging Plastic bottle
CAS 7440-09-7, 7732-18-5
Physical Form Liquid
Molecular Formula K
Concentration 997 to 1003 μg/mL of K(1+) in 2% HNO3 (20°C)
MDL Number MFCD00133776
Color Colorless
Solubility Miscible with water
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
SMILES [K]
IUPAC Name potassium
Molecular Weight (g/mol) 39.10
PubChem CID 5462222
Quantity 500 mL
ChEBI CHEBI:26216
Formula Weight 39.1
Grade AAS
Show More Show Less
compliance-icons
Product Identifier
  • Potassium atomic absorption standard solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.