missing translation for 'onlineSavingsMsg'
Learn More

Potassium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™

Product Code. 10067143
Click to view available options
Quantity:
125 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
K1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 7440-09-7, 7697-37-2, 7732-18-5
Packaging Triple-leached LDPE bottle
CAS Min % 0.1, 2.0, 97.9
Quantity 125 mL
CAS Max % 0.1, 2.0, 97.9
Certificate of Analysis Claritas PPT™ Certificate of Analysis reports up to 68 elements at ppt levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Molecular Formula K
MDL Number MFCD00133776
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
SMILES [K]
IUPAC Name potassium
Molecular Weight (g/mol) 39.10
PubChem CID 5462222
ChEBI CHEBI:26216
Grade Trace ICP/ICP-MS
Solvent or Matrix 2% HNO3
Chemical Name or Material Potassium Standard
Show More Show Less
compliance-icons
Product Identifier
  • 1000
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
Supplemental information
  • MIXTURE LIST-Contains : nitric acid

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'