missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sulfate, 99+%, for analysis, anhydrous, granular, Thermo Scientific Chemicals
30.40 € - 86.30 €
Chemical Identifiers
CAS | 7778-80-5 |
---|---|
Molecular Formula | K2O4S |
Molecular Weight (g/mol) | 174.25 |
MDL Number | MFCD00011388 |
InChI Key | OTYBMLCTZGSZBG-UHFFFAOYSA-L |
Synonym | potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt |
PubChem CID | 24507 |
ChEBI | CHEBI:32036 |
SMILES | [K+].[K+].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10593181
|
Thermo Scientific Acros
205945000 |
500 g | Plastic bottle |
30.40 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10732135
|
Thermo Scientific Acros
205940025 |
2.5 kg | Plastic bottle |
86.30 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisChemische Identifikatoren
7778-80-5 | |
174.25 | |
OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
24507 | |
[K+].[K+].[O-]S([O-])(=O)=O |
K2O4S | |
MFCD00011388 | |
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
CHEBI:32036 |
Spezifikation
1689.0°C | |
7778-80-5 | |
5.5 to 8.5 (5% soln. at 25°C) | |
99+% | |
K2SO4 | |
15, 7793 | |
Solubility in water: 110g/L (20°C). Other solubilities: insoluble in alcohol | |
[K+].[K+].[O-]S([O-])(=O)=O | |
174.25 | |
CHEBI:32036 | |
≥99% | |
2.66 |
1067.0°C | |
White | |
Granular Powder | |
K2O4S | |
MFCD00011388 | |
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
dipotassium;sulfate | |
24507 | |
174.25 | |
Analytical | |
Potassium sulfate, p.a. |
Sicherheit und Handhabung
missing translation for 'einecsNumber' : 231-915-5
missing translation for 'rtecsNumber' : TT5900000
missing translation for 'tsca' : TSCA
RUO – Research Use Only