missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Potassium sulfate, 99+%, for biochemistry Product Code.: 12664997

Thermo Scientific Chemicals Potassium sulfate, 99+%, for biochemistry

Product Code. 12664997
1 kg, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 12664997

Brand: Thermo Scientific Chemicals 424210010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689.0°C
Melting Point 1067.0°C
Color White
Physical Form Crystals
Quantity 1 kg
Assay Percent Range 99+%
Linear Formula K2SO4
Merck Index 15,7793
Solubility Information (10 g in 150 ml water) Clear colorless
Formula Weight 174.25
Percent Purity ≥99%
Grade Biochemical
Packaging Plastic bottle
Density 2.33
Chemical Name or Material Potassium sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0432378 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number424210010,424212500
Lot NumberA0450788 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number424210010,424212500
Lot NumberA0398617 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number424210010,424212500
Lot NumberA0393974 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number424210010,424212500
Lot NumberA0393338 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number424210010,424212500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Potassium sulfate, 99+%, for biochemistry >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.