missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfate, ACS, 99.0% min, Thermo Scientific Chemicals

41.81 € - 121.61 €

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11347877
SDS View Documents
Thermo Scientific Alfa Aesar
014311.36
500 g
41.81 €
500g
Please to purchase this item. Need a web account? Register with us today!
11357877
SDS View Documents
Thermo Scientific Alfa Aesar
014311.A3
2 kg
121.61 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium sulfate is used in the production of fertilizers (i.e. providing potassium and sulfur), potassium alum, potassium carbonate, and glass. In analytical chemistry, it is used in the determination of nitrogen by Kjeldahl method and protein in foodstuffs. Also, it is used in the electrostatic effects on pKa values of amino acid residues in the staphylococcal nuclease. It is used in the separation of protein through electrophoresis and also serves as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7778-80-5
174.25
OTYBMLCTZGSZBG-UHFFFAOYSA-L
24507
[K+].[K+].[O-]S([O-])(=O)=O
K2O4S
MFCD00011388
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
CHEBI:32036

Specifications

1689°C
7778-80-5
99.0% min
MFCD00011388
Soluble in water. Insoluble in alcoholSoluble in water and glycerol. Insoluble in alcohol,acetate and carbon disulfide.
[K+].[K+].[O-]S([O-])(=O)=O
174.25
CHEBI:32036
ACS Reagent
Potassium sulfate, For ACS analysis
1069°C
Crystalline
K2O4S
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium;sulfate
24507
174.27
2.662 g/mL
Videos
Safety and Handling

Safety and Handling

missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III

missing translation for 'einecsNumber' : 231-915-5

missing translation for 'rtecsNumber' : TT5900000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.