missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sulfate, meets analytical spec. of Ph. Eur., 99-101%, Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
46.63 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 kg
50 kg
6 x 1 kg
Packaging:
Fiberboard Box
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 kg |
Linear Formula | K2SO4 |
UN Number | NONH for all modes of transport |
Solubility Information | Insoluble in alcohol, Soluble in boiling water 1gm/4mL, Soluble in glycerol 1gm/75mL, Soluble in water 1gm/8.3mL |
IUPAC Name | dipotassium;sulfate |
Formula Weight | 174.26g/mol |
Percent Purity | 99 to 101% |
Packaging | Poly bottle |
Chemical Name or Material | Potassium Sulfate, Meets analytical specification of Ph.Eur. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'