missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sulfate, pure, Thermo Scientific Chemicals
52.16 € - 401.00 €
Chemical Identifiers
CAS | 7778-80-5 |
---|---|
Molecular Formula | K2O4S |
Molecular Weight (g/mol) | 174.25 |
MDL Number | MFCD00011388 |
InChI Key | OTYBMLCTZGSZBG-UHFFFAOYSA-L |
Synonym | potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt |
PubChem CID | 24507 |
ChEBI | CHEBI:32036 |
SMILES | [K+].[K+].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11954241
|
Thermo Scientific Acros
193810010 |
1 kg | Plastic bottle |
52.16 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11964241
|
Thermo Scientific Acros
193810050 |
5 kg | Plastic bottle |
131.66 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974241
|
Thermo Scientific Acros
193810250 |
25 kg | Fibre drum |
401.00 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7778-80-5 | |
174.25 | |
OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
24507 | |
[K+].[K+].[O-]S([O-])(=O)=O |
K2O4S | |
MFCD00011388 | |
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
CHEBI:32036 |
Specifications
1689.0°C | |
7778-80-5 | |
5.5 to 8.5 | |
K2O4S | |
MFCD00011388 | |
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
dipotassium sulfate | |
24507 | |
174.25 | |
Pure | |
Potassium sulfate |
1067.0°C | |
White | |
Crystalline Powder | |
K2SO4 | |
15,7793 | |
Solubility in water: 110g/L (20°C). Other solubilities: insoluble in alcohol | |
[K+].[K+].[O-]S([O-])(=O)=O | |
174.25 | |
CHEBI:32036 | |
≥99.0% | |
1% max. (130°C) |
Safety and Handling
missing translation for 'einecsNumber' : 231-915-5
RUO – Research Use Only