missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium superoxide, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 400°C |
Color | Yellow |
Physical Form | Chunks and/or Powder |
Quantity | 25 g |
Linear Formula | KO2 |
Solubility Information | Solubility in water: reacts |
Formula Weight | 71.1 |
Packaging | Plastic bottle |
Chemical Name or Material | Potassium superoxide |
Safety and Handling
Product Identifier
- Potassium superoxide
- Danger
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 A
- H272-May intensify fire; oxidiser.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- MIXTURE LIST-Contains : Potassium superoxide
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'