missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-butoxide, 1M solution in tert-butanol, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10030564
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-47-4
Molecular Formula C4H9KO
Molecular Weight (g/mol) 112.21
MDL Number MFCD00012162
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
PubChem CID 23665647
IUPAC Name potassium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[K+]

Specifications

Quantity 100 mL
Linear Formula (CH3)3COK
Flash Point 12°C
Formula Weight 112.21
Concentration or Composition (by Analyte or Components) 0.90 to 1.10M
Packaging AcroSeal™ Glass Bottle
Appearance Colorless to amber solution
Specific Gravity 0.82
Density 0.8200g/mL
Chemical Name or Material Potassium tert-butoxide, 1M solution in tert-butanol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium tert-butoxide, 1M solution in tert-butanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'