missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals

33.27 € - 589.94 €

Chemical Identifiers

CAS 865-47-4
Molecular Formula C4H9KO
Molecular Weight (g/mol) 112.21
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
PubChem CID 23665647
IUPAC Name potassium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[K+]
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10696372
View Documents
Thermo Scientific Acros
168880050
5 g Glass Bottle
33.27 €
5g
Please to purchase this item. Need a web account? Register with us today!
10713504
View Documents
Thermo Scientific Acros
168880250
25 g Plastic bottle
37.59 €
25g
Please to purchase this item. Need a web account? Register with us today!
10419100
View Documents
Thermo Scientific Acros
168881000
100 g Plastic bottle
43.22 €
100g
Please to purchase this item. Need a web account? Register with us today!
10626762
View Documents
Thermo Scientific Acros
168885000
500 g Plastic bottle
140.70 €
500g
Please to purchase this item. Need a web account? Register with us today!
11914611
View Documents
Thermo Scientific Acros
168880025
2.5 kg Plastic bottle
589.94 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

865-47-4
112.21
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
potassium;2-methylpropan-2-olate
C4H9KO
LPNYRYFBWFDTMA-UHFFFAOYSA-N
23665647
CC(C)(C)[O-].[K+]

Specifications

865-47-4
White
C4H9KO
01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289
Solubility in water: reacts with water. Other solubilities: soluble in methanol, ethanol, slightly soluble in diethyl ether, benzene
CC(C)(C)[O-].[K+]
112.21
112.21
Pure
Potassium tert-butoxide
240.0°C to 258.0°C
98+%
(CH3)3COK
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
LPNYRYFBWFDTMA-UHFFFAOYSA-N
potassium;2-methylpropan-2-olate
23665647
98+%
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Self-heating in large quantities; may catch fire.
Reacts violently with water.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several min

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 212-740-3

Product Identifier
  • Potassium tert-butoxide

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1
  • Self-heating substance Category 2
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H228-Flammable solid.
  • H252-Self-heating in large quantities; may catch fire.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only