missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-butoxide, pure, 1.6-1.7M (20 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals

213.22 € - 928.62 €

Specifications

Chemical Name or Material Potassium tert-butoxide
Name Note Pure, 1.7M (20 wt.%) solution in THF
CAS 865-47-4, 109-99-9
Color Amber to Yellow
Physical Form Solution
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10008364
View Documents
Thermo Scientific Acros
428791000
100 mL AcroSeal™ Glass Bottle
213.22 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10287484
View Documents
Thermo Scientific Acros
428798000
800 mL AcroSeal™ Glass Bottle
928.62 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium tert-butoxide, pure, 1.6-1.7M (20 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.