missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-butoxide, pure, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

203.01 € - 2103.00 €

Specifications

Chemical Name or Material Potassium tert-butoxide
CAS 865-47-4, 109-99-9
Color Colorless to Orange-Yellow
Physical Form Solution
Linear Formula (CH3)3COK
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10527003
View Documents Promotion Details
Thermo Scientific Acros
371221000
100 mL AcroSeal™ Glass bottle
203.01 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10436583
View Documents Promotion Details
Thermo Scientific Acros
371220010
1 L AcroSeal™ Glass Bottle
1130.63 €
1 L
Please to purchase this item. Need a web account? Register with us today!
11975851
View Documents Promotion Details
Thermo Scientific Acros
371220025
2.5 L AcroSeal™ Glass Bottle
2103.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium tert-butoxide
Colorless to Orange-Yellow
(CH3)3COK
−19°C
LPNYRYFBWFDTMA-UHFFFAOYSA-N
potassium;2-methylpropan-2-olate
112.21
0.95 to 1.10M
0.9020g/mL
865-47-4, 109-99-9
Solution
01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
CC(C)(C)[O-].[K+]
23665647
Pure
0.902
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
May form explosive peroxides.
Highly flammable liquid and vapor.
Suspected of causing cancer.
Reacts violently with water.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 212-740-3

Product Identifier
  • Potassium tert-butoxide, 1M solution in tetrahydrofuran

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only