Learn More
WTW™ Potassium Test Kits
Brand:  WTW™ 250407
Product Code. 11791113
  • 171.00 € / Pack of 25
Explore more special offers
Specifications
Chromosulfuric Acid oxidation
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex, Photoflex photometers
25
For drinking water, ground water, surface water, well water, curative water, soils
Certificates
Photometric
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • potassium cell test method
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P313-Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethylenedinitrilotetraacetic acid, tetrasodium salt