Learn More
Potassium tetrachloroplatinate, 99.99%, (trace metal basis), Thermo Scientific Chemicals
165.00 € - 567.00 €
Chemical Identifiers
CAS | 10025-99-7 |
---|---|
Molecular Formula | Cl4H4K2Pt |
Molecular Weight (g/mol) | 419.11 |
MDL Number | MFCD00011378 |
InChI Key | JCAXKWJYKUEOFO-UHFFFAOYSA-N |
Synonym | potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate |
PubChem CID | 61440 |
IUPAC Name | dipotassium tetrachloroplatinumdiuide |
SMILES | Cl.Cl.Cl.Cl.[K+].[K+].[Pt--] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11386315
|
Thermo Scientific Acros
437030010 |
1 g | Glass bottle |
165.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11396315
|
Thermo Scientific Acros
437030050 |
5 g | Glass bottle |
567.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
10025-99-7 | |
419.11 | |
JCAXKWJYKUEOFO-UHFFFAOYSA-N | |
61440 | |
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--] |
Cl4H4K2Pt | |
MFCD00011378 | |
potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate | |
dipotassium tetrachloroplatinumdiuide |
Specifications
250.0°C | |
Brown or Orange to Red | |
99.99% | |
K2PtCl4 | |
15,7801 | |
Solubility in water: 10g/L (20°C) | |
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--] | |
419.11 | |
415.1 | |
Trace Metal Basis |
10025-99-7 | |
Crystals | |
Cl4H4K2Pt | |
MFCD00011378 | |
potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate | |
JCAXKWJYKUEOFO-UHFFFAOYSA-N | |
dipotassium tetrachloroplatinumdiuide | |
61440 | |
99.99% | |
Potassium tetrachloroplatinate |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-050-9
RUO – Research Use Only