missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tetrafluoroborate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as additive in alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as additive in alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.
Solubility
Slightly soluble in water and hot ethyl alcohol. Insoluble in cold ethyl alcohol.
Notes
Moisture sensitive. Incompatible withstrong oxidizing agents.
Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as additive in alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.
Solubility
Slightly soluble in water and hot ethyl alcohol. Insoluble in cold ethyl alcohol.
Notes
Moisture sensitive. Incompatible withstrong oxidizing agents.
Specifications
Specifications
Boiling Point | (decomposition) |
Melting Point | 529°C |
Quantity | 5000 g |
Linear Formula | KBF4 |
Merck Index | 14,7686 |
Solubility Information | Slightly soluble in water and hot ethyl alcohol. Insoluble in cold ethyl alcohol. |
Formula Weight | 125.91 |
Percent Purity | 98% |
Refractive Index | 1.3245 |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'