missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrafluoroborate, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10503801
Click to view available options
Quantity:
25 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14075-53-7
Molecular Formula BF4K
Molecular Weight (g/mol) 125.90
MDL Number MFCD00011395
InChI Key AKEBROIVCDHVSD-UHFFFAOYSA-N
Synonym potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride
PubChem CID 518872
IUPAC Name potassium tetrafluoroboranuide
SMILES [K+].F[B-](F)(F)F

Specifications

Melting Point 530.0°C
Color White to Gray
Physical Form Micro-crystalline Powder
Quantity 1 kg
Assay Percent Range 98.5% min
Linear Formula KBF4
Merck Index 15,7804
Solubility Information Solubility in water: 4.4g/L (20°C). Other solubilities: slightly soluble in boiling alcohol
Formula Weight 125.91
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Density 2.5
Chemical Name or Material Potassium tetrafluoroborate, c.p.
Show More Show Less
Product Identifier
  • Potassium tetrafluoroborate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'