missing translation for 'onlineSavingsMsg'
Learn More
Potassium thiobenzoate, 95%, Thermo Scientific Chemicals Product Code.: 15408308

Potassium thiobenzoate, 95%, Thermo Scientific Chemicals

Product Code. 15408308
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15408308

Brand: Thermo Scientific Alfa Aesar H61741.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium thiobenzoate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium thiobenzoate is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28170-13-0
Molecular Formula C7H6KOS
Molecular Weight (g/mol) 177.28
MDL Number MFCD28399074
InChI Key LSBMSRUYNGEWKI-UHFFFAOYSA-N
Synonym benzenecarbothioic acid potassium
PubChem CID 87087528
IUPAC Name benzenecarbothioic S-acid;potassium
SMILES [K].SC(=O)C1=CC=CC=C1

Specifications

Physical Form Solid
Quantity 250 mg
Formula Weight 176.28
Percent Purity 95%
Chemical Name or Material Potassium thiobenzoate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5002Q18W Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberH61741.MD,H61741.03
Lot Number81401351 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61741.MD,H61741.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium thiobenzoate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.