missing translation for 'onlineSavingsMsg'
Learn More
Potassium thiocyanate, 98%, pure, Thermo Scientific Chemicals Product Code.: 10461533

Potassium thiocyanate, 98%, pure, Thermo Scientific Chemicals

Product Code. 10461533
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10461533

Brand: Thermo Scientific Acros 377435000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.18
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium saltShow MoreShow Less
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Melting Point 170.0°C to 179.0°C
Color Colorless or White
Physical Form Crystals
Quantity 500 g
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula KSCN
Merck Index 15,7807
Solubility Information Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 97.18
Percent Purity 98%
Grade Pure
Packaging Plastic bottle
Specific Gravity 1.886
Density 1.8860g/mL
Chemical Name or Material Potassium thiocyanate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464861 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number377435000,377431000,377430025
Lot NumberA0459532 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog Number377435000,377431000,377430025
Lot NumberA0457195 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number377435000,377431000,377430025
Lot NumberA0456498 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number377435000,377431000,377430025
Lot NumberA0450678 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number377435000,377431000,377430025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium thiocyanate, 98%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.