missing translation for 'onlineSavingsMsg'
Learn More
Potassium thiocyanate, 98%, Thermo Scientific Chemicals Product Code.: 11414754

Potassium thiocyanate, 98%, Thermo Scientific Chemicals

Product Code. 11414754
2500 g
Click to view available options
Quantity:
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11414754

Brand: Thermo Scientific Alfa Aesar A13731.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.

Solubility
Soluble in water, acetone and alcohol.

Notes
Hygroscopic. Air, light and moisture sensitive. Incompatible with nitric acids, active halogen compounds and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Melting Point 172°C to 175°C
Density 1.89
pH 5.3 to 8.5
Boiling Point 500°C (decomposition)
Odor Odorless
Quantity 2500 g
Beilstein 3594799
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,7691
Solubility Information Soluble in water,acetone and alcohol.
Formula Weight 97.18
Percent Purity 98%
Chemical Name or Material Potassium thiocyanate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241490 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberA13731.0E,A13731.36
Lot Number10248353 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog NumberA13731.0E,A13731.36
Lot Number10241444 Certificate TypeCertificate of Analysis Date23/06/2022 Catalog NumberA13731.0E,A13731.36
Lot Number10241385 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog NumberA13731.0E,A13731.36
Lot Number10240631 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberA13731.0E,A13731.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium thiocyanate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.