missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium thiocyanate solution, Honeywell™
Click to view available options
Quantity:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
CAS | 333-20-0 |
Quantity | 1 L |
Molecular Formula | CKNS |
Linear Formula | KSCN |
MDL Number | MFCD00011413 |
UN Number | NONH for all modes of transport |
Beilstein | 3594799 |
Synonym | potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt |
InChI Key | ZNNZYHKDIALBAK-UHFFFAOYSA-M |
SMILES | C(#N)[S-].[K+] |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'