missing translation for 'onlineSavingsMsg'
Learn More

Potassium tri-sec-butylborohydride, 1M solution in tetrahydrofuran, AcroSeal™, Thermo Scientific Chemicals

Product Code. p-3779483 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal glass bottle
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10082690

Brand: Thermo Scientific Acros 205471000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium tri-sec-butylborohydride
Name Note 1M Solution in tetrahydrofuran
CAS 54575-49-4, 109-99-9
Color Yellow
Density 0.9100g/mL
Packaging AcroSeal glass bottle
Molecular Formula C12H28BK
Linear Formula KB[CH(CH3)C2H5]3H
MDL Number MFCD00009324
Quantity 100 mL
Fieser 06,490; 07,307
Merck Index 15, 8564
Synonym k-selectride, potassium tri-sec-butylborohydride solution, k-selectride™ solution, k-selectride r solution 1.0 m potassium tri-sec-butylborohydride in thf, k-selectride r solution, 1.0 m potassium tri-sec-butylborohydride in thf, potassium tri-sec-butylborohydride, 1.0m in thf, in sure/seal tm bottle
InChI Key UHNDOQOAVCJJPO-UHFFFAOYSA-N
SMILES [H-].[B+3].[K+].CC[CH-]C.CC[CH-]C.CC[CH-]C
IUPAC Name boron(3+) potassium tris(butan-2-ide) hydride
Molecular Weight (g/mol) 222.26
Formula Weight 222.26
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium tri-sec-butylborohydride, 1 M solution in tetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.