missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium trioxalatoferrate(III) trihydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Description
Potassium trioxalatoferrate(III) trihydrate is used in chemical actinometry to measure light flux.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Green |
Physical Form | Crystalline |
Quantity | 25 g |
UN Number | UN3288 |
Solubility Information | Slightly soluble in water. |
SMILES | C(=O)(C(=O)O)O.C(=O)(C(=O)[O-])O.C(=O)(C(=O)[O-])[O-].[K+].[K+].[K+].[Fe] |
IUPAC Name | tripotassium;2-hydroxy-2-oxoacetate;iron;oxalate;oxalic acid |
Formula Weight | 491.26 (437.21 Anhydrous) |
Sensitivity | Hygroscopic |
Density | 2.133 g/mL |
Show More |
Safety and Handling
Product Identifier
- Potassium trioxalatoferrate(III) trihydrate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Potassium trioxalatoferrate(III) trihydrate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'