Learn More
Potassium tungstate, 99.95%, for trace metal analysis, -100 mesh, Thermo Scientific Chemicals
24.82 € - 68.44 €
Chemical Identifiers
CAS | 7790-60-5 |
---|---|
Molecular Formula | K2O4W |
Molecular Weight (g/mol) | 326.05 |
InChI Key | AAQNGTNRWPXMPB-UHFFFAOYSA-N |
Synonym | potassium tungstate, unii-t6vrm0ou6v, t6vrm0ou6v, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate vi, tungstate wo42-, dipotassium, t-4, 2k.wo4, potassium tungstate-w |
PubChem CID | 3084032 |
IUPAC Name | dipotassium;dioxido(dioxo)tungsten |
SMILES | [O-][W](=O)(=O)[O-].[K+].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13428688
|
Thermo Scientific Acros
448330050 |
5 g | Glass bottle |
24.82 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13438688
|
Thermo Scientific
448330250 |
25 g | Glass bottle |
68.44 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7790-60-5 | |
326.05 | |
potassium tungstate, unii-t6vrm0ou6v, t6vrm0ou6v, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate vi, tungstate wo42-, dipotassium, t-4, 2k.wo4, potassium tungstate-w | |
dipotassium;dioxido(dioxo)tungsten |
K2O4W | |
AAQNGTNRWPXMPB-UHFFFAOYSA-N | |
3084032 | |
[O-][W](=O)(=O)[O-].[K+].[K+] |
Specifications
921°C | |
99.95% | |
K2WO4 | |
potassium tungstate, unii-t6vrm0ou6v, t6vrm0ou6v, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate vi, tungstate wo42-, dipotassium, t-4, 2k.wo4, potassium tungstate-w | |
[O-][W](=O)(=O)[O-].[K+].[K+] | |
326.05 | |
326.05 | |
Trace Metal |
7790-60-5 | |
K2O4W | |
15,7811 | |
AAQNGTNRWPXMPB-UHFFFAOYSA-N | |
dipotassium;dioxido(dioxo)tungsten | |
3084032 | |
99.95% | |
Potassium tungstate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 232-215-2
- Potassium tungstate
RUO – Research Use Only