Learn More
Labcold™ Powder Coated Steel Pharmacy Refrigerators
Click to view available options
Dimensions (DxWxH) Exterior:
725 x 765 x 1710mm
820 x 720 x 1980mm
Unit Size:
Each
Description
- Microprocessor controlled temperature regulation for accuracy and stability
- Temperature range +2°C to +8°C
- Fan circulated interior
- Stainless steel construction
- Door locks fitted as standard
- Digital temperature display with audio alarm
- Heavy duty lockable castors on larger models
- Optimised air circulation
Specifications
Specifications
| Dimensions (DxWxH) Exterior | 725 x 765 x 1710mm |