missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Powder Trier

Constructed of robust stainless steel, making it suitable for sampling cereals, grains, free flowing powders, granules and even slightly cohesive powders. Sampling Systems™ Powder Trier is ideal for taking a large volume, cross sectional sample from containers, sacks or silos.

Brand:  Sampling Systems™ 1216A-600

 View more versions of this product

Product Code. 15462184

  • 940.00 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made from high quality 316L stainless steel
  • Bright finish
  • Suitable for taking a large volume sample from a compacted powder
  • Can take a cross sectional (composite) sample
Specifications

Specifications

Stainless Steel (316L)
110 mL
600 mm
3 Slots
25 mm (excluding scroll)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.