missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ PowderDispo Disposable Sampler
BIOPHARMAGREENER_CHOICE

Designed to sample cross-sections of powdery or slightly cohesive substances. The Buerkle™ PowderDispo Disposable Sampler can sample from bags or similar containers with its lance. Sterile and non-sterile varieties are available.

Brand:  Buerkle™ 5393-5511

154.86 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15541741

  • 182.00 € / Pack of 20
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ideal disposable sampler for cross-section samples of powdery or slightly cohesive substances
  • Complete all-layers samples can be taken from bags or smaller containers with the lance
  • Clean room manufactured
  • Sterile varieties are sterilized by gamma rays
Specifications

Specifications

Sampler
Not Autoclavable
60 mL
Sterile
300 mm
HDPE
2.03 oz.
Single-use
21 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.