Learn More
Power Blotter Welcome Pack, PVDF
The Invitrogen Power Blotter System (Power Blotter Station + Power Blotter Cassette) is designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes.
Brand:  Invitrogen PB0116
2211.00 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 17106039
  • 3350.00 € / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Power Blotter System features an integrated power supply optimized to enable consistent and high-efficiency protein transfer.
The Power Blotter Welcome Pack, PVDF, includes:
  • Power Blotter Station
  • Power Blotter Cassette
  • Power Blotter Select Transfer Stacks, PVDF, regular size
  • Power Blotter Pre-cut Membranes and Filters, PVDF, regular size
  • Power Blotter 1-Step Transfer Buffer
Features of the Power Blotter System include:
  • Efficient—high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
  • Fast—5- to 12-minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
  • Integrated power supply—seamless operation between control unit and cassette; no need for external power supply
  • Easy-touch programming—convenient pre-programmed transfer methods, or create, save, and run customized transfer methods
  • Flexible—simultaneous transfer of 1 or 2 mini-size or 1 midi-size gel using the Power Blotter Cassette
  • Versatile—compatible with Power Blotter Select Transfer Stacks and Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods
The Power Blotter System achieves highly efficient transfer in seven minutes when used with Power Blotter Select Transfer Stacks or ten minutes with Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional buffers.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Acrylamide Gels
PVDF
Room Temperature
Power Blotter
1 Kit
• Power Blotter Station
• Power Blotter Cassette
• Power Blotter Select Transfer Stacks, PVDF, regular-size
• Power Blotter Pre-cut Membranes and Filters, PVDF, regular-size
• Power Blotter 1-Step Transfer Buffer
Semi-Dry