missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PowerEase™ Touch Power Supply

Invitrogen PowerEase Touch power supplies are user-friendly and easy to use with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the pre-programmed gel electrophoresis and transfer methods a breeze.

Brand:  Invitrogen™ PS0121

683.40 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 16630396

  • 830.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Includes the power supply and a quick reference card.
Gel Electrophoresis
500 mA
Power Supply
1 each
300 V
PowerEase Touch 120W
DNA/RNA Electrophoresis, SDS-PAGE, and Native PAGE
PowerEase Touch
230 VAC
Room Temperature
120 W
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ PowerEase™ Touch Power Supply > PowerEase Touch 120W, 230 VAC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.