Learn More
Ansell™ Powerflex™ 80-400 Series Natural Rubber Latex Heavy Weight Gloves Product Code.: 11776673
Add to basket
Ansell™ Powerflex™ 80-400 Series Natural Rubber Latex Heavy Weight Gloves
Product Code. 11776673 Shop All Ansell Products
10
Click to view available options
Size:
7
8
10
11
Product Code. 11776673
Brand: Ansell™ 80400100
Add to basket
Add to basket
Provide outstanding insulation from cold conditions, as well as high comfort and grip. Combines a thermal terry loop acrylic liner with a naturally insulating latex coating, ensuring excellent insulation against the cold.
  • Provides comfort and protection for the wearer in temperatures down to as low as -30°C, for short or intermittent contact.
  • Terry loop acrylic liner that is  in color
  • Bright yellow liner profices extra high visibility in low or poor light conditions for additional safety
  • Natural insulating latex coating
  • Knitted construction
  • Palm-coated finish
  • Knitwrist cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material Rubber Latex
Certifications/Compliance Standard CATEGORY III EN 388 : X231, EN 407 : X2XXXX, EN 511 : 020, ANSI
Cuff Material Natural Rubber Latex
Coating Material Natural Rubber Latex
Product Type Heavyweight Gloves
Size 10
Style 80-400
Glove Cuff Style Knit
Lining Acrylic terry loop
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To Grip, abrasion and puncture resistance
Series 80-400
Product Title
Ansell™ Powerflex™ 80-400 Series Natural Rubber Latex Heavy Weight Gloves >