Learn More
Ansell™ PowerFlex™ 80-658 Series Aramid Fiber Heavy Weight Gloves
Protects wearer with triple-layer construction comprised of steel, glass fiber and aramid fiber provides extreme resistance to cuts, splinters and burrs.
323.00 €
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11726723
|
Ansell™
80658100 |
10 |
323.00 €
12 pairs |
In Stock
Log in to see stock. |
|||||
12647011
|
Ansell™
80658110 |
11 |
323.00 €
12 pairs |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
12657011
|
Ansell™
80658060 |
6 |
323.00 €
12 pairs |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
11725834
|
Ansell™
80658080 |
8 |
323.00 €
12 pairs |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
11736723
|
Ansell™
80658090 |
9 |
323.00 €
12 pairs |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
15646906
|
Ansell™
80658070 |
7 |
323.00 €
12 pairs |
In Stock
Log in to see stock. |
|||||
Description
- Mechanical protection gloves
- Exceeds EN388 cut level 5.
- Extra-long cuff offers extended safety for the forearm.
- ¾ dipped and liquid repellent, shielding the skin from unpleasant liquid solutions.
- Good heat contact resistance.
- EN407 level-2 rating means safe handling of warm objects.
- Natural rubber crinkle finish provides a firm grip.
- Long life provides a highly cost-effective solution.
- Heather green color helps mask dirt.
- Excellent washability, with no decrease in comfort or fading.
Applications
- Handling heavy loads, metal edges
- Waste handling: recycling, sorting (glass, cans, metal pieces)
- Handling glass sheets, sharp-edged objects
- Handling metal sheets, cutting of dry, painted or galvanized pieces
Spécification
No | |
Long Knitwrist |