Learn More
3M™ Powerflow™ XL Air Flow Indicator 031-34-02P
3M™ Powerflow™ XL Air Flow Indicator 031-34-02P is used to confirm that the minimum design flow rate of the 3M™ Powerflow™ Powered Air Respirator is exceeded.
Brand:  3M™ 7000104091
Product Code. 15729897
  • 4.75 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Air Flow indicator is used to confirm that the manufacturer's minimum design flow rate is exceeded
  • Easy to use
  • Genuine 3M™ Accessory
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3M™ Powerflow™ Powered Air Respirator