Learn More
WLD Tec™ Powerjet 2 Buckling Gun
Brand:  WLD Tec™ 6.006.000
Product Code. 10367492
  • 583.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Specifications
Liquid Gas, Natural Gas
Microbiology or Biotechnical Fields
1300°C (Natural Gas), 1350°C (Liquid Gas)
150 mm
150 mm
210 mm
500 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Manuals
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
WLD Tec™ Powerjet 2 Buckling Gun > Height: 210mm