Learn More
Fisherbrand™ PowerPro 3 Amp
Fisherbrand™ PowerPro 3 Amp is a power supply designed for virtually all high current electrophoresis applications.
Brand:  Fisherbrand™ FPOWERPLUS3AMP
Product Code. 15828481
  • 1335.00 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 5 pairs of outputs
  • Higher current output capability is perfect for electroblotting units with high-intensity plate electrodes, particularly Cleaver Scientific omniBLOT maxi, VS20 WAVE and semi dry blotting systems.
  • Electrotransfers may be performed as timed runs in constant or programmable mode to prevent overheating and buffer depletion.
  • A run time extendable to a maximum 999 minutes in constant mode is also useful for overnight transfers undertaken at constant low current in wire electrode systems.
  • Shares the built-in protocol and constant parameter features of the other PowerPro models.
Specifications
Specifications
Electroblotting units with high-intensity plate electrodes
2.4 in. TFT-LCD
300 w output, 1 w increments
Constant mode: 9999 (min.) with alarm Programmable mode: 999 (min.) with alarm
100 to 240 V rated, 5 to 300 V output, 1 V increments
10 to 3000 mA output, 10 mA increments