Learn More
Fisherbrand™ PowerPro 500
Fisherbrand™ PowerPro 500 is an excellent general-purpose power supply that fits the broadest range of electrophoresis applications, and can run as many as four units under a constant or programmable setting.
Brand:  Fisherbrand™ FPOWERPLUS500
Product Code. 15838481
  • 1705.00 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Perfect for horizontal or vertical electrophoresis
  • Rigorous in-built safety mechanisms
  • Dual voltage compatibility
  • 5 pairs of outlets
  • It has the capacity to store up to 30 programmed files, each with 6 steps.
  • Perfect for techniques that benefit from subtle and gradual stepwise changes in the electrical parameters as the run progresses, such as DGGE and large format vertical PAGE using Cleaver Scientific VS20-DGGE and WAVE systems.
  • Built-in pre-set protocols from running each of Cleaver Scientific’s electrophoresis systems
  • Users can create and save protocols for their own systems.
  • A 2.4” TFT-LCD Screen displays the current running parameters and allows the user to browse pre-set and saved protocols.
  • Can be operated at constant voltage, constant current or constant power. The convenient pause/resume key allows any run to be temporarily halted for inspection of the gel and restarted from the same position.
Specifications
Specifications
Broad range of electrophoresis applications
2.4 in. TFT-LCD
300 w output, 1 w increments
Constant mode: 9999 (min.) with alarm Programmable mode: 999 (min.) with alarm
100 to 240 V rated, 5 to 500 V output, 1 V increments
1 to 800 mA output, 1 mA increments