Learn More
Bel-Art™ Scienceware™ Poxy Coated Wire Framed Bag Holder
Collects pipette tips, microcentrifuge tubes. Bel Art™ Poxy Coated Wire Framed Bag Holder is economical and convenient.
Brand: Bel-Art™ Scienceware™ H13193-1000
This item is not returnable.
View return policy
Description
Collects pipette tips, microcentrifuge tubes. Bel Art™ Poxy Coated Wire Framed Bag Holder is economical and convenient.
- Rectangular steel wire frame holds a polyethylene bag 22 x 28cm (8½ x 11 in.); coated in easy to clean, durable, Orange color epoxy
- Snug fitting polypropylene plastic cover helps hold the bag securely in place
- Reusable; can be cleaned by hand or in automated dish washers; autoclavable.
- Cover provides compliance with 29 CFR 1910.1030(d)(2)(xiii)(A), which requires a cover on any container used to store, transport, or ship biohazardous wastes
- Can be used with Bench Top Biohazard Bags (F13166-0000); can also be used with Clavies Autoclavable Bags (H13182-0812)
Specifications
Bag Holder | |
Orange | |
For bench top biohazard bags | |
Epoxy-coated Wire |