Learn More
Bel-Art™ Poxygrid™ Biohazard Bag Holders
Sturdy wire holder keeps bags open while a separate hooked ring that rests over the top keeps them securely in place
Brand: Bel-Art™ H13188-0000
Description
Sturdy wire holder keeps bags open while a separate hooked ring that rests over the top keeps them securely in place
- Bright biohazard orange colored epoxy-coated steel wire
- Chemical and corrosion-resistant; unaffected by organic solvents, salts, weak acids and weak alkalis
- Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F)
- Autoclavable
Specifications
Biohazard Bag Holder | |
Autoclavable | |
Reusable | |
01-814-2, 01-814C, 01-815B, 01-826-6, 01-826B, 01-827C, 01-830D, 01-831A, 01-828-033, 01-832A 30 H x 24 in. W biohazard bags | |
-369°F to +250°F | |
14.6 in. | |
23.5 in. | |
14.6 in. |
Circular epoxy coated wire frame with easy-to-use hooked ring over a top | |
Orange | |
To keep biohazard bags open and ready for use | |
Epoxy-coated Wire | |
-223°C to +121°C | |
37 cm | |
60 cm | |
37 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.