missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Poxygrid™ Wire Test Tube Racks
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
Brand: Bel-Art™ F18760-1150
This item is not returnable.
View return policy
Description
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
- Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
- Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
- Orange racks can be used for biohazard warning while yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Stores from 40 to 108 tubes (five different diameters of testtubes)
- Autoclavable
Specifications
Test Tube Rack | |
Autoclavable | |
Tubes (13 to 16 mm) | |
150 | |
3 Tiers | |
Non-sterile | |
305 mm | |
209 mm | |
64 mm | |
150 mm |
10 x 15 | |
Blue | |
Epoxy-coated Steel | |
Rectangular | |
Yes, When Empty | |
12 x 8.25 x 2.5 in. (30.5 x 21 x 6.4cm) | |
12 in. | |
8.25 in. | |
2.5 in. | |
16 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction