missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Poxygrid™ Wire Test Tube Racks

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

Brand:  Bel-Art™ F18759-0000

 View more versions of this product

Product Code. 15560022

  • 54.81 € / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

  • Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
  • Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
  • Orange racks can be used for biohazard warning while yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Stores from 40 to 108 tubes (five different diameters of testtubes)
  • Autoclavable
Specifications

Specifications

Test Tube Rack
Autoclavable
Tubes (13 to 16 mm)
108
3 Tiers
Non-sterile
242 mm
189 mm
64 mm
16 mm
9 x 12
Green
Epoxy-coated Steel
Rectangular
Yes, When Empty
9.5 x 7.4 x 2.5 in. (242 x 189 x 64mm)
9.48 in.
7.4 in.
2.51 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.