Learn More
RETSCH PP 25 Pellet Press
Prepare solid samples for XRF analysis with this pellet press. Retsch™ PP 25 Pellet Press is a compact benchtop unit with particularly simple and safe operation. Produces high quality pellets characterized by their high degree of stability.
Brand:  RETSCH 20.750.0006
Product Code. 11924572
Description
Description
  • Produces high quality, stable pellets
  • Easy and safe operation
  • Several pressing tools available
  • Can be evacuated
  • Compact benchtop unit
Specifications
Specifications
Pellet Press
Polypropylene
0.8 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
RETSCH PP 25 Pellet Press > Pellet Press