missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP5C Mouse anti-Canine, Human, Clone: OTI4G8, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500592S
This item is not returnable.
View return policy
Description
PPP5C Monoclonal antibody specifically detects PPP5C in Canine, Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| PPP5C | |
| Monoclonal | |
| 0.9 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P53041 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5536, 612199 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4G8 | |
| Unconjugated | |
| Ppp5c | |
| ANP receptor; AU020526; EGK_10788; FLJ36922; FLJ55954; OTTHUMP00000165834; PP5; PPP5; Ppp5c; PPT; PP-T; protein phosphatase 5 catalytic subunit; protein phosphatase 5, catalytic subunit; protein phosphatase T; Serine/threonine-protein phosphatase 5 | |
| Full length human recombinant protein of human PPP5C produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction