Learn More
Grant Instruments™ PQ Documentation
Offers equipment user greater comparability and verification with their instruments. Grant Instruments™ PQ Documentation simplifies installation and maintenance procedures, reduces common user errors, and builds a clear record of an instrument's performance throughout its life.
Brand:  Grant Instruments™ PQ QBH
 View more versions of this product
Product Code. 15211216
  • 348.00 € / Each
Explore more special offers
Description
Description
Performance Qualification (PQ): Instrument is continuing to meet its specification
  • Each documentation set is specific to individual serial numbered equipment and is provided in an easy to use ring bound format.
  • PQ documentation can be ordered at any time.
Specifications
Specifications
PQ Documentation